missing translation for 'onlineSavingsMsg'
Learn More

GE Healthcare Dharmacon DharmaFECT™-1 siRNA Transfection Reagent

Most broadly applicable DharmaFECT formulation for optimal siRNA or microRNA transfection into a wide range of cell types for successful RNAi experiments

Brand:  GE Healthcare Dharmacon T-2001-01

Code : P1

Additional Details : Weight : 0.00752kg

 View more versions of this product

Product Code. 11571731

  • £73.26 / 0.20mL


Estimated Shipment
14-03-2018
Add to basket

Specifications

Specifications

1mg/mL
High efficiency of siRNA and microRNA delivery with minimal impact on cell viability across a broad range of cell types for successful RNAi experiments
DharmaFECT-1 Reagent
0.2mL
Videos