missing translation for 'onlineSavingsMsg'
Learn More

GE Healthcare Dharmacon DharmaFECT™-2 siRNA Transfection Reagent

Most broadly applicable DharmaFECT formulation for optimal siRNA or microRNA transfection into a wide range of cell types for successful RNAi experiments

Brand:  GE Healthcare Dharmacon T-2002-02

Code : P1

Additional Details : Weight : 1.27460kg

 View more versions of this product

Product Code. 11521741

  • £210.90 / 0.75mL


Estimated Shipment
08-02-2018
Add to basket

Description

Description

  • Optimized for small RNA (siRNA or microRNA) delivery
  • DharmaFECT provides four specialized lipid-based formulations for successful delivery to more cell lines than any other single reagent
  • DharmaFECT 1 is the most all-purpose transfection reagent, demonstrating efficient, low-toxicity delivery to over 80% of validated cell types
  • DharmaFECT 2, 3, and 4 offer distinct formulations to support a wider range of cell types, and permit more thorough optimization of transfection for high-value experiments and screening projects

Specifications

Specifications

1mg/mL
High efficiency of siRNA and microRNA delivery with minimal impact on cell viability across a broad range of cell types for successful RNAi experiments
DharmaFECT-2 Reagent
0.75mL
Videos