Learn More
GE Healthcare Dharmacon DharmaFECT™-2 siRNA Transfection Reagent
Most broadly applicable DharmaFECT formulation for optimal siRNA or microRNA transfection into a wide range of cell types for successful RNAi experiments
Brand:  GE Healthcare Dharmacon T-2002-01
Code : P1
Additional Details : Weight : 1.27460kg
 View more versions of this product
Product Code. 11511741
  • £73.26 / 0.20mL
Estimated Shipment date
from Supplier 23-01-2018
Add to basket
Description
Description
  • Optimized for small RNA (siRNA or microRNA) delivery
  • DharmaFECT provides four specialized lipid-based formulations for successful delivery to more cell lines than any other single reagent
  • DharmaFECT 1 is the most all-purpose transfection reagent, demonstrating efficient, low-toxicity delivery to over 80% of validated cell types
  • DharmaFECT 2, 3, and 4 offer distinct formulations to support a wider range of cell types, and permit more thorough optimization of transfection for high-value experiments and screening projects
Specifications
Specifications
1mg/mL
High efficiency of siRNA and microRNA delivery with minimal impact on cell viability across a broad range of cell types for successful RNAi experiments
DharmaFECT-2 Reagent
0.2mL
Videos