missing translation for 'onlineSavingsMsg'
Learn More

GE Healthcare Dharmacon DharmaFECT™-4 siRNA Transfection Reagent

Most broadly applicable DharmaFECT formulation for optimal siRNA or microRNA transfection into a wide range of cell types for successful RNAi experiments

Brand:  GE Healthcare Dharmacon T-2004-01

UNSPSC : 41106402

Code : P1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11591741

  • £73.26 / 0.20mL


Estimated Shipment
04-07-2017
Add to basket

Specifications Description & Specifications

Specifications

1mg/mL
DharmaFECT-4 Reagent
High efficiency of siRNA and microRNA delivery with minimal impact on cell viability across a broad range of cell types for successful RNAi experiments
0.2mL