Learn More
GE Healthcare Dharmacon DharmaFECT™-3 siRNA Transfection Reagent
Most broadly applicable DharmaFECT formulation for optimal siRNA or microRNA transfection into a wide range of cell types for successful RNAi experiments
Brand:  GE Healthcare Dharmacon T-2003-04
UNSPSC : 41116133
Code : P1
Additional Details : Weight : 0.30000kg
 View more versions of this product
Product Code. 12169740
  • £1656.12 / Pack of 5
Estimated Shipment date
from Supplier 29-06-2017
Add to basket
Specifications Description & Specifications
Specifications
1mg/mL
High efficiency of siRNA and microRNA delivery with minimal impact on cell viability across a broad range of cell types for successful RNAi experiments
DharmaFECT-3 Reagent
5 x 1.5mL