missing translation for 'onlineSavingsMsg'
Learn More

GE Healthcare Dharmacon DharmaFECT™-3 siRNA Transfection Reagent

Most broadly applicable DharmaFECT formulation for optimal siRNA or microRNA transfection into a wide range of cell types for successful RNAi experiments

Brand:  GE Healthcare Dharmacon T-2003-03

UNSPSC : 41106402

Code : P1

Additional Details : Weight : 1.27460kg

 View more versions of this product

Product Code. 11571741

  • £351.50 / 1.50mL


Estimated Shipment date
from Supplier 06-07-2017
Add to basket

Specifications Description & Specifications

Specifications

1mg/mL
High efficiency of siRNA and microRNA delivery with minimal impact on cell viability across a broad range of cell types for successful RNAi experiments
DharmaFECT-3 Reagent
1.5mL