GE Healthcare Dharmacon DharmaFECT™-3 siRNA Transfection Reagent
Most broadly applicable DharmaFECT formulation for optimal siRNA or microRNA transfection into a wide range of cell types for successful RNAi experiments
Overview
Brand: GE Healthcare Dharmacon
Manufacturer Part Number: T-2003-02
DHARMAFECT 3 0,75ML
UNSPSC: 41106402
Code: P1
Additional Details:
Additional Details: Weight: 1.27460kg
Product Code. 11561741
Quantity Price
1 £ 210.9 / 0.75mL
Estimated Shipment date
from Supplier 01-03-2017
Add to basket
Description and Specification
Specification
Concentration 1mg/mL
Product Type DharmaFECT-3 Reagent
Concentration 1mg/mL
Quantity 0.75mL
For Use With (Application) High efficiency of siRNA and microRNA delivery with minimal impact on cell viability across a broad range of cell types for successful RNAi experiments
Quantity 0.75mL