GE Healthcare Dharmacon DharmaFECT™-3 siRNA Transfection Reagent

Most broadly applicable DharmaFECT formulation for optimal siRNA or microRNA transfection into a wide range of cell types for successful RNAi experiments

Overview
Brand: GE Healthcare Dharmacon

Manufacturer Part Number: T-2003-01

UNSPSC: 41106402

Code: P1

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 11551741

Quantity Price
1 £ 73.26 / 0.20mL
Estimated Shipment date
from Supplier 04-05-2017
Add to basket


Description and Specification

Specification

Concentration 1mg/mL
For Use With (Application) High efficiency of siRNA and microRNA delivery with minimal impact on cell viability across a broad range of cell types for successful RNAi experiments
Product Type DharmaFECT-3 Reagent
Quantity 0.2mL