GE Healthcare Dharmacon DharmaFECT™ siRNA Transfection Reagents Set (1-4)
Most broadly applicable DharmaFECT formulation for optimal siRNA or microRNA transfection into a wide range of cell types for successful RNAi experiments
Overview
Brand: GE Healthcare Dharmacon
Manufacturer Part Number: T-2005-02
DHARMAFECT 1, 2, 3, 4 4PIECES
UNSPSC: 12352200
Code: P1
Additional Details:
Additional Details: Weight: 0.01000kg
Product Code. 11687330
Quantity Price
1 £ 805.12 / Pack of 4
Estimated Shipment date
from Supplier 26-01-2017
Add to basket
Description and Specification
Specification
Concentration 1mg/mL
Product Type DharmaFECT Set (1-4)
Concentration 1mg/mL
Quantity 4 x 0.75mL
For Use With (Application) High efficiency of siRNA and microRNA delivery with minimal impact on cell viability across a broad range of cell types for successful RNAi experiments
Quantity 4 x 0.75mL