GE Healthcare Dharmacon DharmaFECT™ Duo Transfection Reagents

Reagents optimized for delivery of both siRNA and plasmid in a single formulation

Overview
Brand: GE Healthcare Dharmacon

Manufacturer Part Number: T-2010-04

UNSPSC: 41116133

Code: P1

Additional Details:
Additional Details: Weight: 0.30000kg



Product Code. 12179760

Quantity Price
1 £ 1656.12 / Pack of 5
Estimated Shipment date
from Supplier 03-04-2017
Add to basket


Description and Specification

Specification

Concentration 1mg/mL
Concentration 1mg/mL
Quantity 5 x 1.5mL
For Use With (Application) Delivery of both siRNA and plasmid in a single formulation for RNAi screening and reporter systems
Quantity 5 x 1.5mL