GE Healthcare Dharmacon DharmaFECT™ Duo Transfection Reagents

Reagents optimized for delivery of both siRNA and plasmid in a single formulation

Overview
Brand: GE Healthcare Dharmacon

Manufacturer Part Number: T-2010-02

UNSPSC: 41106402

Code: P1

Additional Details:
Additional Details: Weight: 1.27460kg



Product Code. 11571751

Quantity Price
1 £ 210.9 / 0.75mL
Estimated Shipment date
from Supplier 03-05-2017
Add to basket


Description and Specification

Specification

Concentration 1mg/mL
For Use With (Application) Delivery of both siRNA and plasmid in a single formulation for RNAi screening and reporter systems
Quantity 0.75mL