GE Healthcare Dharmacon DharmaFECT™ Duo Transfection Reagents

Reagents optimized for delivery of both siRNA and plasmid in a single formulation

Overview
Brand: GE Healthcare Dharmacon

Manufacturer Part Number: T-2010-03

UNSPSC: 12000000

Code: P1

Additional Details:
Additional Details: Weight: 1.27460kg



Product Code. 10425374

Quantity Price
1 £ 351.5 / 1.50mL
Estimated Shipment date
from Supplier 31-03-2017
Add to basket


Description and Specification

Specification

Concentration 1mg/mL
For Use With (Application) Delivery of both siRNA and plasmid in a single formulation for RNAi screening and reporter systems
Quantity 1.5mL