Learn More
GE Healthcare Dharmacon Accell™ siRNA Positive Control Reagents
non-species-specific controls for targeting of reporter genes or other experimental needs
Brand:  GE Healthcare Dharmacon D-002050-01-20
UNSPSC : 41106402
Code : P1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11591711
  • £214.60 / 20nmol
Estimated Shipment date
from Supplier 26-09-2017
Add to basket
Specifications
Specifications
Effectively silences the pGL3 luciferase reporter, use in any mammalian cell type
siRNA Positive Control
20nm