Learn More
GE Healthcare Dharmacon Accell™ siRNA Positive Control Reagents
non-species-specific controls for targeting of reporter genes or other experimental needs
Brand:  GE Healthcare Dharmacon D-002000-01-20
UNSPSC : 41106402
Code : P1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11581711
  • £214.60 / 20nmol
Estimated Shipment date
from Supplier 28-06-2017
Add to basket
Specifications Description & Specifications
Specifications
siRNA Positive Control
Validated control for efficient silencing of beta-galactosidase, use in any mammalian cell type
20nm