GE Healthcare Dharmacon Accell™ siRNA Positive Control Reagents
non-species-specific controls for targeting of reporter genes or other experimental needs
Overview
Brand: GE Healthcare Dharmacon
Manufacturer Part Number: D-001940-10-50
UNSPSC: 41106402
Code: P1
Additional Details:
Additional Details: Weight: 0.01000kg
Product Code. 11551671
Quantity Price
1 £ 667.48 / 50nmol
Estimated Shipment date
from Supplier 08-05-2017
Add to basket
Description and Specification
Specification
Product Type siRNA Positive Control
For Use With (Application) Accell™ siRNA enters cells without the need for transfection reagents, virus (or viral vectors) or instruments, targets over 100 known eGFP cloning vectors and transgenic sequences
Quantity 50nm