Learn More
GE Healthcare Dharmacon Accell™ siRNA Positive Control Reagents
non-species-specific controls for targeting of reporter genes or other experimental needs
Brand:  GE Healthcare Dharmacon D-001940-10-05
Code : P1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11531671
  • £168.72 / 5nmol
Call for latest availability
+44 (0)1509 555 500
Add to basket
Specifications
Specifications
Accell™ siRNA enters cells without the need for transfection reagents, virus (or viral vectors) or instruments, targets over 100 known eGFP cloning vectors and transgenic sequences
siRNA Positive Control
5nm
Videos