Learn More
GE Healthcare Dharmacon Accell™ siRNA Delivery Media
Specially formulated and optimized for Accell siRNA delivery
Brand:  GE Healthcare Dharmacon B-005000-500
UNSPSC : 41106402
Code : P1
Additional Details : Weight : 3.17510kg
 View more versions of this product
Product Code. 11577170
  • £27.38 / 500mL
Estimated Shipment date
from Supplier 02-06-2017
Add to basket
Specifications Description & Specifications
Specifications
500mL
Accell™ siRNA reagents which provides appropriate Serum-free conditions for Accell™ uptake while maintaining cell health