Accell™ Nontargeting Control siRNA and Nontargeting Pool
Designed to aid in the interpretation of experiments with Accell siRNA reagents as highly reliable controls for assessment of baseline cellular responses
Overview
Brand: GE Healthcare Dharmacon
Manufacturer Part Number: D-001910-03-50
ACCELL NONTARG SIR 3 50NMOL
UNSPSC: 41106402
Code: P1
Additional Details:
Additional Details: Weight: 0.01000kg
Product Code. 11577270
Quantity Price
1 £ 616.42 / 50nmol
Estimated Shipment date
from Supplier 29-03-2017
Add to basket
Description and Specification
Specification
Product Type Nontargeting siRNA No. 3 Control siRNA
Quantity 50nm
For Use With (Application) Designed to aid in the interpretation of experiments with Accell™ siRNA reagents as highly reliable controls for assessment of baseline cellular responses
Quantity 50nm