Learn More
GE Healthcare Dharmacon Accell™ Nontargeting Control siRNA and Nontargeting Pool
Designed to aid in the interpretation of experiments with Accell siRNA reagents as highly reliable controls for assessment of baseline cellular responses
Brand:  GE Healthcare Dharmacon D-001910-02-20
Code : P1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11537270
  • £364.82 / 20nmol
Call for latest availability
+44 (0)1509 555 500
Add to basket
Specifications
Specifications
Designed to aid in the interpretation of experiments with Accell™ siRNA reagents as highly reliable controls for assessment of baseline cellular responses
Nontargeting siRNA No. 2 Control siRNA
20nm
Videos