Learn More
GE Healthcare Dharmacon Accell™ Nontargeting Control siRNA and Nontargeting Pool
Designed to aid in the interpretation of experiments with Accell siRNA reagents as highly reliable controls for assessment of baseline cellular responses
Brand:  GE Healthcare Dharmacon D-001910-10-50
UNSPSC : 41106402
Code : P1
Additional Details : Weight : 0.22680kg
 View more versions of this product
Product Code. 11537280
  • £667.48 / 50nmol
Estimated Shipment date
from Supplier 31-07-2017
Add to basket
Specifications Description & Specifications
Specifications
Designed to aid in the interpretation of experiments with Accell™ siRNA reagents as highly reliable controls for assessment of baseline cellular responses
Nontargeting Pool
50nm