Learn More
GE Healthcare Dharmacon Accell™ Nontargeting Control siRNA and Nontargeting Pool
Designed to aid in the interpretation of experiments with Accell siRNA reagents as highly reliable controls for assessment of baseline cellular responses
Brand:  GE Healthcare Dharmacon D-001910-10-20
Code : P1
Additional Details : Weight : 0.22680kg
 View more versions of this product
Product Code. 11527280
  • £394.42 / 20nmol
Estimated Shipment
08-02-2018
Add to basket
Specifications
Specifications
Designed to aid in the interpretation of experiments with Accell™ siRNA reagents as highly reliable controls for assessment of baseline cellular responses
Nontargeting Pool
20nm
Videos