Learn More
GE Healthcare Dharmacon Accell™ GAPD Control siRNA and GAPD Pool
Recommended as a positive silencing control
Brand:  GE Healthcare Dharmacon D-001930-10-50
UNSPSC : 41106402
Code : P1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11531661
  • £667.48 / 50nmol
Estimated Shipment date
from Supplier 03-07-2017
Add to basket
Specifications Description & Specifications
Specifications
NM_002046
Designed to aid in the interpretation of experiments with Accell™ siRNA reagents as highly reliable positive controls for delivery and silencing capability
GAPD Pool—Human
50nm