GE Healthcare Dharmacon Accell™ GAPD Control siRNA and GAPD Pool
Recommended as a positive silencing control
Overview
Brand: GE Healthcare Dharmacon
Manufacturer Part Number: D-001930-10-20
UNSPSC: 41106402
Code: P1
Additional Details:
Additional Details: Weight: 0.22680kg
Product Code. 11521661
Quantity Price
1 £ 394.42 / 20nmol
Estimated Shipment date
from Supplier 08-05-2017
Add to basket
Description and Specification
Specification
Product Type GAPD Pool—Human
Accession Number NM_002046
Quantity 20nm
For Use With (Application) Designed to aid in the interpretation of experiments with Accell™ siRNA reagents as highly reliable positive controls for delivery and silencing capability