Accell™ GAPD Control siRNA and GAPD Pool
Recommended as a positive silencing control
Overview
Brand: GE Healthcare Dharmacon
Manufacturer Part Number: D-001930-03-50
UNSPSC: 41106402
Code: P1
Additional Details:
Additional Details: Weight: 0.01000kg
Product Code. 11501661
Quantity Price
1 £ 421.06 / 50nmol
Estimated Shipment date
from Supplier 01-06-2017
Add to basket
Description and Specification
Specification
Accession Number XM_575242
For Use With (Application) Designed to aid in the interpretation of experiments with Accell™ siRNA reagents as highly reliable positive controls for delivery and silencing capability
Product Type GAPD Control siRNA—Rat
Quantity 50nm