GE Healthcare Dharmacon Accell™ GAPD Control siRNA and GAPD Pool
Recommended as a positive silencing control
Overview
Brand: GE Healthcare Dharmacon
Manufacturer Part Number: D-001930-30-50
ACCELL GAPD POOL-RAT 50NMOL
UNSPSC: 41106402
Code: P1
Additional Details:
Additional Details: Weight: 0.01000kg
Product Code. 11591661
Quantity Price
1 £ 667.48 / 50nmol
Estimated Shipment date
from Supplier 27-02-2017
Add to basket
Description and Specification
Specification
Accession Number XM_575242
Product Type GAPD Pool—Rat
Quantity 50nm
For Use With (Application) Designed to aid in the interpretation of experiments with Accell™ siRNA reagents as highly reliable positive controls for delivery and silencing capability
Quantity 50nm