GE Healthcare Dharmacon Accell™ GAPD Control siRNA and GAPD Pool
Recommended as a positive silencing control
Overview
Brand: GE Healthcare Dharmacon
Manufacturer Part Number: D-001930-02-05
UNSPSC: 41106402
Code: P1
Additional Details:
Additional Details: Weight: 0.01000kg
Product Code. 11551651
Quantity Price
1 £ 114.7 / 5nmol
Estimated Shipment date
from Supplier 05-06-2017
Add to basket
Description and Specification
Specification
Accession Number NM_008085
For Use With (Application) Designed to aid in the interpretation of experiments with Accell™ siRNA reagents as highly reliable positive controls for delivery and silencing capability
Product Type GAPD Control siRNA—Mouse
Quantity 5nm