GE Healthcare Dharmacon Accell™ Fluorescent Nontargeting Control siRNA
Provides highly reliable qualitative assessment of Accell siRNA uptake as determined by fluorescent microscopy to evaluate cytoplamic localization of the dye-labeled Accell siRNA FACS analysis to determine general uptake
Overview
Brand: GE Healthcare Dharmacon
Manufacturer Part Number: D-001950-01-50
ACCELL G NONTARG SIR(FAM) 50NMOL
UNSPSC: 41106402
Code: P1
Additional Details:
Additional Details: Weight: 0.01000kg
Product Code. 11581671
Quantity Price
1 £ 799.94 / 50nmol
Estimated Shipment date
from Supplier 20-02-2017
Add to basket
Description and Specification
Specification
Fluorophore FITC
Quantity 50nm
Filter Type FITC
For Use With (Application) Evaluate cytoplamic localization of the dye-labeled Accell™ siRNA FACS analysis to determine general uptake and provides highly reliable qualitative assessment of Accell™ siRNA uptake as determined by Fluorescent micros
Quantity 50nm