Learn More
GE Healthcare Dharmacon Accell™ Fluorescent Nontargeting Control siRNA
Provides highly reliable qualitative assessment of Accell siRNA uptake as determined by fluorescent microscopy to evaluate cytoplamic localization of the dye-labeled Accell siRNA FACS analysis to determine general uptake
Brand:  GE Healthcare Dharmacon D-001950-01-05
UNSPSC : 41106402
Code : P1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11561671
  • £171.68 / 5nmol
Estimated Shipment date
from Supplier 29-06-2017
Add to basket
Specifications Description & Specifications
Specifications
FITC
Evaluate cytoplamic localization of the dye-labeled Accell™ siRNA FACS analysis to determine general uptake and provides highly reliable qualitative assessment of Accell™ siRNA uptake as determined by Fluorescent micros
5nm