GE Healthcare Dharmacon Accell™ Fluorescent Nontargeting Control siRNA
Provides highly reliable qualitative assessment of Accell siRNA uptake as determined by fluorescent microscopy to evaluate cytoplamic localization of the dye-labeled Accell siRNA FACS analysis to determine general uptake
Overview
Brand: GE Healthcare Dharmacon
Manufacturer Part Number: D-001960-01-50
UNSPSC: 41106402
Code: P1
Additional Details:
Additional Details: Weight: 0.01000kg
Product Code. 11511681
Quantity Price
1 £ 799.94 / 50nmol
Estimated Shipment date
from Supplier 08-05-2017
Add to basket
Description and Specification
Specification
Fluorophore Cy3 Thodamine or Polyethylene
For Use With (Application) Evaluate cytoplamic localization of the dye-labeled Accell™ siRNA FACS analysis to determine general uptake and provides highly reliable qualitative assessment of Accell™ siRNA uptake as determined by Fluorescent micros
Quantity 50nm