GE Healthcare Dharmacon Accell™ Fluorescent Cyclophilin B Control siRNA

Provides highly reliable qualitative assessment of Accell siRNA uptake as determined by fluorescent microscopy to evaluate cytoplamic localization of the dye-labeled Accell siRNA FACS analysis to determine general uptake

Overview
Brand: GE Healthcare Dharmacon

Manufacturer Part Number: D-001970-02-05

ACCELL G CYCLO SIR-MO(FAM) 5NMOL

UNSPSC: 41106402

Code: P1

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 11551681

Quantity Price
1 £ 171.68 / 5nmol
Estimated Shipment date
from Supplier 24-02-2017
Add to basket


Description and Specification

Specification

Accession Number NM_011149
Fluorophore FITC
Product Type Green (FAM) Cyclophilin B Control siRNA
Color Green
Species Mouse
Quantity 5nmol
Filter Type FITC
For Use With (Application) Highly reliable qualitative assessment of Accell™ siRNA uptake as determined by Fluorescent microscopy to evaluate cytoplamic localization of the dye-labeled Accell siRNA FACS analysis to determine general uptake
Quantity 5nmol