Learn More
GE Healthcare Dharmacon Accell™ Fluorescent Cyclophilin B Control siRNA
Provides highly reliable qualitative assessment of Accell siRNA uptake as determined by fluorescent microscopy to evaluate cytoplamic localization of the dye-labeled Accell siRNA FACS analysis to determine general uptake
Brand:  GE Healthcare Dharmacon D-001970-02-20
UNSPSC : 41106402
Code : P1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11561681
  • £473.60 / 20nmol
Estimated Shipment date
from Supplier 31-07-2017
Add to basket
Specifications Description & Specifications
Specifications
NM_011149
FITC
Green (FAM) Cyclophilin B Control siRNA
Mouse
Green
Highly reliable qualitative assessment of Accell™ siRNA uptake as determined by Fluorescent microscopy to evaluate cytoplamic localization of the dye-labeled Accell siRNA FACS analysis to determine general uptake
20nmol