Accell™ Fluorescent Cyclophilin B Control siRNA

Provides highly reliable qualitative assessment of Accell siRNA uptake as determined by fluorescent microscopy to evaluate cytoplamic localization of the dye-labeled Accell siRNA FACS analysis to determine general uptake

Overview
Brand: GE Healthcare Dharmacon

Manufacturer Part Number: D-001975-01-50

UNSPSC: 41106402

Code: P1

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 11531691

Quantity Price
1 £ 799.94 / 50nmol
Estimated Shipment date
from Supplier 02-06-2017
Add to basket


Description and Specification

Specification

Accession Number NM_000942
Color Red
Fluorophore Cy3 Thodamine or Polyethylene
For Use With (Application) Highly reliable qualitative assessment of Accell™ siRNA uptake as determined by Fluorescent microscopy to evaluate cytoplamic localization of the dye-labeled Accell siRNA FACS analysis to determine general uptake
Product Type Red (DY-547) Cyclophilin B Control siRNA
Quantity 50nmol
Species Human