GE Healthcare Dharmacon Accell™ eGFP Control siRNA
Silences expression of the GFP protein, leading to a detectable lack of fluorescence as observed by fluorescent microscopy or by FACS analysis
Overview
Brand: GE Healthcare Dharmacon
Manufacturer Part Number: D-001940-01-20
ACCELL EGFP SIR 20NMOL
UNSPSC: 12352200
Code: P1
Additional Details:
Additional Details: Weight: 0.80000kg
Product Code. 11812074
Quantity Price
1 £ 252.34 / 20nmol
Estimated Shipment date
from Supplier 31-03-2017
Add to basket
Description and Specification
Specification
For Use With (Application) Designed to aid in the interpretation of experiments with Accell™ siRNA reagents as highly reliable positive controls for delivery and silencing capability
Quantity 20nm
Quantity 20nm