Learn More
GE Healthcare Dharmacon Accell™ eGFP Control siRNA
Silences expression of the GFP protein, leading to a detectable lack of fluorescence as observed by fluorescent microscopy or by FACS analysis
Brand:  GE Healthcare Dharmacon D-001940-01-50
UNSPSC : 41106402
Code : P1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11521671
  • £421.06 / 50nmol
Estimated Shipment date
from Supplier 29-08-2017
Add to basket
Specifications Description & Specifications
Specifications
Designed to aid in the interpretation of experiments with Accell™ siRNA reagents as highly reliable positive controls for delivery and silencing capability
50nm