Learn More
GE Healthcare Dharmacon Accell™ eGFP Control siRNA
Silences expression of the GFP protein, leading to a detectable lack of fluorescence as observed by fluorescent microscopy or by FACS analysis
Brand:  GE Healthcare Dharmacon D-001940-01-05
Code : P1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11501671
  • £114.70 / 5nmol
Estimated Shipment date
from Supplier 21-02-2018
Add to basket
Specifications
Specifications
Designed to aid in the interpretation of experiments with Accell™ siRNA reagents as highly reliable positive controls for delivery and silencing capability
5nm
Videos