Learn More
GE Healthcare Dharmacon Accell™ Cyclophilin B Control siRNA and Cyclophilin B Pool
Recommended as a positive silencing control
Brand:  GE Healthcare Dharmacon D-001920-10-05
Code : P1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11531641
  • £168.72 / 5nmol
Call for latest availability
+44 (0)1509 555 500
Add to basket
Specifications
Specifications
NM_000942
Designed to aid in the interpretation of experiments with Accell™ siRNA reagents as highly reliable positive controls for delivery and silencing capability
Cyclophilin B Pool—Human
5nm
Videos