Learn More
GE Healthcare Dharmacon Accell™ Cyclophilin B Control siRNA and Cyclophilin B Pool
Recommended as a positive silencing control
Brand:  GE Healthcare Dharmacon D-001920-20-05
UNSPSC : 41106402
Code : P1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11561641
  • £168.72 / 5nmol
Estimated Shipment date
from Supplier 28-06-2017
Add to basket
Specifications Description & Specifications
Specifications
NM_011149
Designed to aid in the interpretation of experiments with Accell™ siRNA reagents as highly reliable positive controls for delivery and silencing capability
Cyclophilin B Pool—Mouse
5nm