Accell™ Cyclophilin B Control siRNA and Cyclophilin B Pool
Recommended as a positive silencing control
Overview
Brand: GE Healthcare Dharmacon
Manufacturer Part Number: D-001920-30-50
UNSPSC: 41106402
Code: P1
Additional Details:
Additional Details: Weight: 0.01000kg
Product Code. 11511651
Quantity Price
1 £ 667.48 / 50nmol
Estimated Shipment date
from Supplier 10-05-2017
Add to basket
Description and Specification
Specification
Accession Number NM_022536
Product Type Cyclophilin B Pool—Rat
For Use With (Application) Designed to aid in the interpretation of experiments with Accell™ siRNA reagents as highly reliable positive controls for delivery and silencing capability
Quantity 50nm