Learn More
Accell™ Cyclophilin B Control siRNA and Cyclophilin B Pool
Recommended as a positive silencing control
Brand:  GE Healthcare Dharmacon D-001920-30-20
UNSPSC : 41106402
Code : P1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11501651
  • £394.42 / 20nmol
Estimated Shipment date
from Supplier 31-07-2017
Add to basket
Specifications Description & Specifications
Specifications
NM_022536
Designed to aid in the interpretation of experiments with Accell™ siRNA reagents as highly reliable positive controls for delivery and silencing capability
Cyclophilin B Pool—Rat
20nm