Learn More
GE Healthcare Dharmacon Accell™ Control siRNA Kits
Accell siRNA represents a breakthrough technology to empower RNAi, without transfection reagents, viral vectors, or instrumentation
Brand:  GE Healthcare Dharmacon K-005000-G1-03
UNSPSC : 12352200
Code : P1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11652710
  • £413.66 / Each
Estimated Shipment date
from Supplier 30-08-2017
Add to basket
Specifications Description & Specifications
Specifications
Accell siRNA Control Kit (Green)
Rat
5nM Tubes of each of the four validated control siRNAs (Accell™ non-targeting siRNA #1, Accell™ Cyclophilin B siRNA and GAPD siRNA, Accell™ green non-targeting siRNA), 5X siRNA Buffer, 100mL delivery m
Green
Each
Assessing Accell™ technology in a given cell type and to ensure effective controls during initial gene silencing experiments with Accell™ siRNA reagents