GE Healthcare Dharmacon Accell™ Control siRNA Kits

Accell siRNA represents a breakthrough technology to empower RNAi, without transfection reagents, viral vectors, or instrumentation

Overview
Brand: GE Healthcare Dharmacon

Manufacturer Part Number: K-005000-R1-02

UNSPSC: 12352200

Code: P1

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 11622710

Quantity Price
1 £ 413.66 / Each
Estimated Shipment date
from Supplier 02-05-2017
Add to basket


Description and Specification

Specification

Product Type Accell siRNA Control Kit (Red)
Color Red
Species Mouse
Includes 5nM Tubes of each of the four validated control siRNAs (Accell™ non-targeting siRNA #1, Accell™ Cyclophilin B siRNA and GAPD siRNA, Accell™ red non-targeting siRNA), 5X siRNA Buffer, 100mL delivery med
For Use With (Application) Assessing Accell™ technology in a given cell type and to ensure effective controls during initial gene silencing experiments with Accell™ siRNA reagents
Quantity Each