GE Healthcare Dharmacon Accell™ Control siRNA Kits

Accell siRNA represents a breakthrough technology to empower RNAi, without transfection reagents, viral vectors, or instrumentation

Overview
Brand: GE Healthcare Dharmacon

Manufacturer Part Number: K-005000-R1-01

UNSPSC: 12352200

Code: P1

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 11612710

Quantity Price
1 £ 413.66 / Each
Estimated Shipment date
from Supplier 08-05-2017
Add to basket


Description and Specification

Specification

Product Type Accell siRNA Control Kit (Red)
Color Red
Species Human
For Use With (Application) Assessing Accell™ technology in a given cell type and to ensure effective controls during initial gene silencing experiments with Accell™ siRNA reagents
Includes 5nM Tubes of each of the four validated control siRNAs (Accell™ non-targeting siRNA #1, Accell™ Cyclophilin B siRNA and GAPD siRNA, Accell™ red non-targeting siRNA), 5X siRNA Buffer, 100mL delivery med
Quantity Each