missing translation for 'onlineSavingsMsg'
Learn More

GE Healthcare cAMP Direct BioTrak™ EIA

Use for direct measurement of cAMP in cell lysates without any need for removal of extraction reagents. GE Healthcare Life Sciences™ cAMP Direct BioTrak™ EIA represents a convenient, high-sensitivity, peroxidase-based technology that enables the amount of protein present in a sample to be measured in a single day.

Brand:  GE Healthcare RPN225

Code : GE

Additional Details : Weight : 2.80320kg Transport : UN number : 3316 Chem class : 1 Pack group : II


 View more versions of this product

Product Code. 10585155

  • £599.20 / Each


Estimated Shipment
21-02-2018
Add to basket

If this item is out of stock, delivery will be in 3-4 working days providing the item is in stock at the supplier.
Please enquire with customer services on 01509 555500 for stock availability.

 

Description

Description

  • Flexible method-choice of assay protocols
  • Rapid assay protocol
  • Non-radioactive
  • Precise and accurate measurement
  • Ready-to-use substrate
  • Color coded reagents
  • Five-hour protocol

  • Kits available with or without acetylation reagents
  • Bulk kit provided without lysis buffers
  • Dual range: 25–6400 fmol/well [non-acetylation], 2–128 fmol/well [acetylation]
  • Sensitivity: 38.4pg/mL [non-acetylation] and 14pg/mL [acetylation]

Specifications

Specifications

Used for research purposes, eliminates lengthy sample extraction procedures and allow direct cAMP measurement
96 Determinations
2° to 8°C
cAMP biotrak Enzyme immunoassay (EIA) system
38.4pg/mL (non-acetylation), 14pg/mL (acetylation)
96-well Microplate
Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 A
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Serious eye damage/eye irritation Category 1
  • Acute toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
  • H360D-May damage the unborn child.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H412-Harmful to aquatic life with long lasting effects.
  • H318-Causes serious eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P235-Keep cool.
  • P501-Dispose of contents/container to .
  • P273-Avoid release to the environment.
  • P270-Do not eat, drink or smoke when using this product.
  • P405-Store locked up.
  • P201-Obtain special instructions before use.
  • P331-Do NOT induce vomiting.

Supplemental information
  • MIXTURE LIST-Contains¬†: calcium chloride