missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Butyl Sepharose™ High Performance Chromatography Media
Designed for intermediate and polishing step purification steps
£117.69 - £692.00
Specifications
For Use With (Application) | For Intermediate and Polishing Step PurifiCatIon Steps |
---|---|
Stability | Stable in commonly used aqueous Buffers: 1 M Sodium Hydroxide, 1 M Acetic Acid, 8 M Urea, 6 M Guanidine Hydrochloride, 30% Acetonitrile, 70% Ethanol, 3 M Ammonium sulfate, 1 mM HCl, 30% Isopropanol, 2% SDS |
Storage Requirements | 4°C to 30°C |
Binding Capacity | 38 mg b-lactoglobulin/mL mediμm |
Product Type | Butyl Sepharose High Performance Media |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10363060
|
Cytiva
17-5432-02 |
200 mL |
£692.00
200mL |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
10640761
|
Cytiva
17-5432-01 |
25 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Hydrophobic interaction chromatography (HIC) media composed of highly cross-linked 34μm agarose beads with aliphatic butyl functional groups
- High-resolution, capacity separations for recovery
- High ligand density for dynamic binding capacity even at low salt concentration
- Display minimum of ionic interactions
- Chemical stability for effective CIP and sanitization
- Available in prepacked columns such as HiPrep™, HiScreen™, and HiTrap™ plus laboratory and BioProcess pack sizes
Specifications
For Intermediate and Polishing Step PurifiCatIon Steps | |
4°C to 30°C | |
Butyl Sepharose High Performance Media | |
24 to 44 μm | |
Cross-Linked Agarose | |
20% Ethanol |
Stable in commonly used aqueous Buffers: 1 M Sodium Hydroxide, 1 M Acetic Acid, 8 M Urea, 6 M Guanidine Hydrochloride, 30% Acetonitrile, 70% Ethanol, 3 M Ammonium sulfate, 1 mM HCl, 30% Isopropanol, 2% SDS | |
38 mg b-lactoglobulin/mL mediμm | |
100 to 200 cm/Hoμr | |
300 kPa | |
3 to 13 | |
Butyl |