Learn More
Cytiva Butyl Sepharose™ High Performance Chromatography Media
Designed for intermediate and polishing step purification steps
Brand: Cytiva 17-5432-01
Code : GE
Additional Details : CAS Number : 64-17-5 Weight : 0.02500kg
Description
- Hydrophobic interaction chromatography (HIC) media composed of highly cross-linked 34μm agarose beads with aliphatic butyl functional groups
- High-resolution, capacity separations for recovery
- High ligand density for dynamic binding capacity even at low salt concentration
- Display minimum of ionic interactions
- Chemical stability for effective CIP and sanitization
- Available in prepacked columns such as HiPrep™, HiScreen™, and HiTrap™ plus laboratory and BioProcess pack sizes
Specifications
25 mL | |
Stable in commonly used aqueous Buffers: 1 M Sodium Hydroxide, 1 M Acetic Acid, 8 M Urea, 6 M Guanidine Hydrochloride, 30% Acetonitrile, 70% Ethanol, 3 M Ammonium sulfate, 1 mM HCl, 30% Isopropanol, 2% SDS | |
38 mg b-lactoglobulin/mL mediμm | |
100 to 200 cm/Hoμr | |
300 kPa | |
3 to 13 | |
Butyl |
For Intermediate and Polishing Step PurifiCatIon Steps | |
4°C to 30°C | |
Butyl Sepharose High Performance Media | |
24 to 44 μm | |
Cross-Linked Agarose | |
20% Ethanol |
Safety and Handling
- Butyl Sepharose High Performance, 25 ml
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P235-Keep cool.
- P501-Dispose of contents/container to .
Supplemental information
- MIXTURE LIST-Contains : ethanol