missing translation for 'onlineSavingsMsg'
Learn More

Life Science products Cytiva Spacer 20mm x 240mm x 1.5mm for SE 400/600

Brand:  Life Science products Cytiva 80619096

Spacer 20mm x 240mm x 1.5mm for SE 400/600

Code : GH

Additional Details : Weight : 0.05000kg

Product Code. 11360442

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.