missing translation for 'onlineSavingsMsg'
Learn More

Life Science products Cytiva Comb 5.7mm x 0.75mm 15 well for SE 400/600

Brand:  Life Science products Cytiva 80616113

Comb 5.7mm x 0.75mm 15 well for SE 400/600

Code : GH

Additional Details : Weight : 0.05000kg

Product Code. 11340502

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.