missing translation for 'onlineSavingsMsg'
Learn More

GE Healthcare Amersham™ Protran™ Supported NC Nitrocellulose Western Blotting Sandwiches

Easy-to-use, precut, preassembled PVDF membrane and filter paper sandwiches

Brand:  GE Healthcare 10600108

Code : GH

Additional Details : Weight : 0.27000kg

 View more versions of this product

Product Code. 15279874

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description

Description

  • Preassembled with two Whatman™ medium thickness 3MM Chr (0.34mm) filter papers to simplify set up
  • Fit most precase gel formats

Specifications

Specifications

14cm
16cm
Sandwich, Pk. of 10
0.45μm
Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Flammable solid Category 1

Hazard Statement
  • H228-Flammable solid.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.