Learn More
GE Healthcare Amersham™ Protran™ Supported NC Nitrocellulose Membranes: Sheets
Provides exceptional handling characteristics, allowing reprobing
Brand:  GE Healthcare 10600098
Code : GH
Additional Details : Weight : 0.08200kg
 View more versions of this product
Product Code. 15299864
  • £84.72 / 25 sheets
Estimated Shipment
08-06-2018
Add to basket
Description
Description
Individual sheets.
  • Membranes consist of nitrocellulose supported by an inert polyester nonwoven material within the membrane
  • The support does not affect transfer conditions or results
Specifications
Specifications
25/Pk.
Nitrocellulose and inert polyester
0.45um
9cm
Sheets
Videos