GE Healthcare Amersham™ Protran™ Premium NC Nitrocellulose Western Blotting Sandwiches

Precut, preassembled Protran Premium NC Nitrocellulose membrane and filter paper sandwiches save time

Overview
Brand: GE Healthcare

Manufacturer Part Number: 10600106

UNSPSC: 41105339

Code: GH

Additional Details:
Additional Details: Weight: 0.25500kg Transport: UN number : 3270 Chem class : 1 Pack group : II



Product Code. 15259874

Quantity Price
1 £ 110.34 / Pack of 10
Estimated Shipment
13-04-2017
Add to basket


Description and Specification

Specification

Format Sandwich, Pk. of 10
Width (Metric) 16cm
Length (Metric) 14cm
Pore Size 0.45μm